missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoethyl Fumarate 97.0+%, TCI America™
$167.49 - $1171.28
Chemical Identifiers
| CAS | 5-4-2459 |
|---|---|
| Molecular Formula | C6H8O4 |
| Molecular Weight (g/mol) | 144.13 |
| MDL Number | MFCD00002699 |
| InChI Key | XLYMOEINVGRTEX-ONEGZZNKSA-N |
| Synonym | monoethyl fumarate, e-4-ethoxy-4-oxobut-2-enoic acid, ethyl hydrogen fumarate, fumaric acid monoethyl ester, e-4-ethoxy-4-oxo-2-butenoic acid, unii-y3w849ng2n, 2e-4-ethoxy-4-oxobut-2-enoic acid, fumaric acid, monoethyl ester, mono-ethyl fumarate, ethyl hydrogen maleate |
| PubChem CID | 5358902 |
| IUPAC Name | (E)-4-ethoxy-4-oxobut-2-enoic acid |
| SMILES | CCOC(=O)C=CC(=O)O |
Chemical Identifiers
| 5-4-2459 | |
| 144.13 | |
| XLYMOEINVGRTEX-ONEGZZNKSA-N | |
| 5358902 | |
| CCOC(=O)C=CC(=O)O |
| C6H8O4 | |
| MFCD00002699 | |
| monoethyl fumarate, e-4-ethoxy-4-oxobut-2-enoic acid, ethyl hydrogen fumarate, fumaric acid monoethyl ester, e-4-ethoxy-4-oxo-2-butenoic acid, unii-y3w849ng2n, 2e-4-ethoxy-4-oxobut-2-enoic acid, fumaric acid, monoethyl ester, mono-ethyl fumarate, ethyl hydrogen maleate | |
| (E)-4-ethoxy-4-oxobut-2-enoic acid |
Specifications
| 5-4-2459 | |
| White | |
| C6H8O4 | |
| 25 g | |
| XLYMOEINVGRTEX-ONEGZZNKSA-N | |
| (E)-4-ethoxy-4-oxobut-2-enoic acid | |
| 5358902 | |
| ≥97.0% (GC,T) | |
| Monoethyl Fumarate |
| 68°C | |
| 147°C | |
| MFCD00002699 | |
| monoethyl fumarate, e-4-ethoxy-4-oxobut-2-enoic acid, ethyl hydrogen fumarate, fumaric acid monoethyl ester, e-4-ethoxy-4-oxo-2-butenoic acid, unii-y3w849ng2n, 2e-4-ethoxy-4-oxobut-2-enoic acid, fumaric acid, monoethyl ester, mono-ethyl fumarate, ethyl hydrogen maleate | |
| CCOC(=O)C=CC(=O)O | |
| 144.13 | |
| 144.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1142
TSCA : No