missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Fumarate 97.0+%, TCI America™

Supplier:  TCI America F035925G

 View more versions of this product

Catalog No. F035925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Monoethyl Fumarate
68°C
147°C
MFCD00002699
monoethyl fumarate, e-4-ethoxy-4-oxobut-2-enoic acid, ethyl hydrogen fumarate, fumaric acid monoethyl ester, e-4-ethoxy-4-oxo-2-butenoic acid, unii-y3w849ng2n, 2e-4-ethoxy-4-oxobut-2-enoic acid, fumaric acid, monoethyl ester, mono-ethyl fumarate, ethyl hydrogen maleate
CCOC(=O)C=CC(=O)O
144.13
144.13
Crystalline Powder
5-4-2459
White
C6H8O4
25 g
XLYMOEINVGRTEX-ONEGZZNKSA-N
(E)-4-ethoxy-4-oxobut-2-enoic acid
5358902
≥97.0% (GC,T)

Chemical Identifiers

5-4-2459
144.13
XLYMOEINVGRTEX-ONEGZZNKSA-N
5358902
CCOC(=O)C=CC(=O)O
C6H8O4
MFCD00002699
monoethyl fumarate, e-4-ethoxy-4-oxobut-2-enoic acid, ethyl hydrogen fumarate, fumaric acid monoethyl ester, e-4-ethoxy-4-oxo-2-butenoic acid, unii-y3w849ng2n, 2e-4-ethoxy-4-oxobut-2-enoic acid, fumaric acid, monoethyl ester, mono-ethyl fumarate, ethyl hydrogen maleate
(E)-4-ethoxy-4-oxobut-2-enoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1142

TSCA : No

SDS
Documents

Documents

Promotions

Promotions