missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Adipate 97.0+%, TCI America™

Catalog No. A122325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A122325G 25 g
A1223500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A122325G Supplier TCI America Supplier No. A122325G
Only null left

Chemical Identifiers

CAS 626-86-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00004419
InChI Key UZNLHJCCGYKCIL-UHFFFAOYSA-N
Synonym adipic acid monoethyl ester, monoethyl adipate, ethyl hydrogen adipate, hexanedioic acid, monoethyl ester, monoethyl hexanedioate, monoethyladipic acid ester, adipic acid, monoethyl ester, mono-ethyl adipate, hexanedioic acid, 1-ethyl ester, hexanoic acid, monoethyl ester
PubChem CID 12295
IUPAC Name 6-ethoxy-6-oxohexanoic acid
SMILES CCOC(=O)CCCCC(=O)O

Specifications

Quantity 25 g
Formula Weight 174.20
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material Monoethyl Adipate
EINECSNumber (2)-0861
RTECSNumber AV2030000
TSCA Yes