accessibility menu, dialog, popup

UserName

H3COOOHO

CAS RN 626-86-8

IUPAC Name: 6-ethoxy-6-oxohexanoic acid
Synonyms: hexanedioic acid, monoethyl ester
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: UZNLHJCCGYKCIL-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCC(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)