missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl 4-Oxoheptanedioate 97.0+%, TCI America™

Catalog No. O02115G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02115G Supplier TCI America Supplier No. O02115G
Only null left

Chemical Identifiers

CAS 1506-55-4
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00059093
InChI Key JYEYJELELKLPLO-UHFFFAOYSA-N
Synonym 4-Oxoheptanedioic Acid Monoethyl Ester
PubChem CID 568249
IUPAC Name 7-ethoxy-4,7-dioxoheptanoic acid
SMILES CCOC(=O)CCC(=O)CCC(O)=O

Specifications

Melting Point 70°C
Color White
Quantity 5 g
Formula Weight 202.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monoethyl 4-Oxoheptanedioate