accessibility menu, dialog, popup

UserName

H3COOOOOH

CAS RN 1506-55-4

IUPAC Name: 7-ethoxy-4,7-dioxoheptanoic acid
Synonyms: 7-ethoxy-4,7-dioxoheptanoic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C9H14O5
InChi Key: JYEYJELELKLPLO-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(=O)CCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results