missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monocaprylin 98.0+%, TCI America™
$141.67 - $559.22
Chemical Identifiers
| CAS | 26402-26-6 |
|---|---|
| Molecular Formula | C11H22O4 |
| Molecular Weight (g/mol) | 218.293 |
| MDL Number | MFCD00056652 |
| InChI Key | GHBFNMLVSPCDGN-UHFFFAOYSA-N |
| Synonym | Glycerol alpha-Monooctanoate, 1-Monooctanoyl Glycerol |
| PubChem CID | 3033877 |
| ChEBI | CHEBI:85241 |
| IUPAC Name | 2,3-dihydroxypropyl octanoate |
| SMILES | CCCCCCCC(=O)OCC(CO)O |
Chemical Identifiers
| 26402-26-6 | |
| 218.293 | |
| GHBFNMLVSPCDGN-UHFFFAOYSA-N | |
| 3033877 | |
| 2,3-dihydroxypropyl octanoate |
| C11H22O4 | |
| MFCD00056652 | |
| Glycerol alpha-Monooctanoate, 1-Monooctanoyl Glycerol | |
| CHEBI:85241 | |
| CCCCCCCC(=O)OCC(CO)O |
Specifications
| 26402-26-6 | |
| White-Yellow | |
| MFCD00056652 | |
| Glycerol alpha-Monooctanoate, 1-Monooctanoyl Glycerol | |
| CCCCCCCC(=O)OCC(CO)O | |
| 218.293 | |
| CHEBI:85241 | |
| ≥98.0% (GC) | |
| Monocaprylin |
| 41°C | |
| C11H22O4 | |
| 1 g | |
| GHBFNMLVSPCDGN-UHFFFAOYSA-N | |
| 2,3-dihydroxypropyl octanoate | |
| 3033877 | |
| 218.29 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0669
TSCA : Yes