accessibility menu, dialog, popup

UserName

H3COOOHOH

CAS RN 26402-26-6

IUPAC Name: 2,3-dihydroxypropyl octanoate
Synonyms: monoctanoin
Molecular Weight (g/mol): 218.29
Molecular Formula: C11H22O4
InChi Key: GHBFNMLVSPCDGN-UHFFFAOYNA-N
SMILES: CCCCCCCC(=O)OCC(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results