missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monobutyl Phthalate 95.0+%, TCI America™
$152.86 - $436.19
Chemical Identifiers
| CAS | 131-70-4 |
|---|---|
| Molecular Formula | C12H14O4 |
| Molecular Weight (g/mol) | 222.24 |
| MDL Number | MFCD00043498 |
| InChI Key | YZBOVSFWWNVKRJ-UHFFFAOYSA-N |
| Synonym | Phthalic Acid Monobutyl Ester, Butyl Hydrogen Phthalate |
| PubChem CID | 8575 |
| IUPAC Name | 2-(butoxycarbonyl)benzoic acid |
| SMILES | CCCCOC(=O)C1=CC=CC=C1C(O)=O |
Chemical Identifiers
| 131-70-4 | |
| 222.24 | |
| YZBOVSFWWNVKRJ-UHFFFAOYSA-N | |
| 8575 | |
| CCCCOC(=O)C1=CC=CC=C1C(O)=O |
| C12H14O4 | |
| MFCD00043498 | |
| Phthalic Acid Monobutyl Ester, Butyl Hydrogen Phthalate | |
| 2-(butoxycarbonyl)benzoic acid |
Specifications
| 131-70-4 | |
| White | |
| MFCD00043498 | |
| Phthalic Acid Monobutyl Ester, Butyl Hydrogen Phthalate | |
| CCCCOC(=O)C1=CC=CC=C1C(O)=O | |
| 222.24 | |
| 222.24 | |
| Crystalline Powder |
| 73°C | |
| C12H14O4 | |
| 5 g | |
| YZBOVSFWWNVKRJ-UHFFFAOYSA-N | |
| 2-(butoxycarbonyl)benzoic acid | |
| 8575 | |
| ≥95.0% (GC,T) | |
| Monobutyl Phthalate |
Safety and Handling
EINECSNumber : (3)-1308
RTECSNumber : TI2475000
TSCA : Yes