missing translation for 'onlineSavingsMsg'
Learn More

Monobutyl Phthalate Copper(II) Salt 95.0+%, TCI America™

Catalog No. P067625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P067625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P067625G Supplier TCI America Supplier No. P067625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25215-53-6
Molecular Formula C24H26CuO8
Molecular Weight (g/mol) 506.01
MDL Number MFCD00045860
InChI Key FQHSBEXCSPGVGY-UHFFFAOYSA-L
Synonym Butyl Phthalate Copper(II) Salt, Phthalic Acid Monobutyl Ester Copper(II) Salt
PubChem CID 79467
IUPAC Name copper;2-butoxycarbonylbenzoate
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)[O-].CCCCOC(=O)C1=CC=CC=C1C(=O)[O-].[Cu+2]

Specifications

Quantity 25 g
Formula Weight 506.01
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monobutyl Phthalate Copper(II) Salt
EINECSNumber (3)-1384
TSCA No