accessibility menu, dialog, popup

UserName

CAS RN 25215-53-6

IUPAC Name: copper(2+) bis(2-(butoxycarbonyl)benzoate)
Synonyms: copper(2+) bis(monobutyl phthalate)
Molecular Weight (g/mol): 506.01
Molecular Formula: C24H26CuO8
InChi Key: FQHSBEXCSPGVGY-UHFFFAOYSA-L
SMILES: [Cu++].CCCCOC(=O)C1=CC=CC=C1C([O-])=O.CCCCOC(=O)C1=CC=CC=C1C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results