missing translation for 'onlineSavingsMsg'
Learn More

Monobutyl Itaconate 95.0+%, TCI America™

Catalog No. I026725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I026725G 25 g
I0267500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I026725G Supplier TCI America Supplier No. I026725G

Chemical Identifiers

CAS 6439-57-2
Molecular Formula C9H13O4-
Molecular Weight (g/mol) 185.199
MDL Number MFCD00046533
InChI Key DYYFCJRYZVHEKQ-UHFFFAOYSA-M
Synonym beta-Butyl Itaconate, 4-Butyl Methylenesuccinate, Itaconic Acid Monobutyl Ester
PubChem CID 21225725
IUPAC Name 3-butoxycarbonylbut-3-enoate
SMILES CCCCOC(=O)C(=C)CC(=O)[O-]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 186.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Monobutyl Itaconate
EINECSNumber (2)-1127
TSCA No