missing translation for 'onlineSavingsMsg'
Learn More

Monobenzyl Phthalate 98.0+%, TCI America™

Catalog No. P119110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P119110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P119110G Supplier TCI America Supplier No. P119110G
Only null left

Chemical Identifiers

CAS 2528-16-7
Molecular Formula C15H11CaO4
Molecular Weight (g/mol) 295.33
MDL Number MFCD00020279
InChI Key HLBMYHRBXPVVCC-UHFFFAOYSA-M
Synonym Benzyl Hydrogen Phthalate, Phthalic Acid Monobenzyl Ester
PubChem CID 31736
IUPAC Name calcium 2-[(benzyloxy)carbonyl]benzoate
SMILES [Ca++].[O-]C(=O)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 106°C
Color White
Quantity 10 g
Formula Weight 256.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monobenzyl Phthalate
RTECSNumber CZ4322500
TSCA Yes