missing translation for 'onlineSavingsMsg'
Learn More

Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP, 98-100.5%, Spectrum™ Chemical

Catalog No. 18604666
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18604666 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604666 Supplier Spectrum Chemical Mfg Cor Supplier No. SO130500GM

SO130, 10049-21-5, NaH2PO4·H2O

Spectrum™ Chemical Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP - All Spectrum Chemical USP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

CAS Max % 1%
pH 4.2 to 4.5
Quantity 500 g
Percent Purity 98 to 100.5%
Grade Multi-Compendial
Loss on Drying 11.5-14.5%
Packaging Poly Bottle