missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monobasic Sodium Phosphate, Dihydrate, USP, 98-103%, Spectrum™ Chemical
S1930, 13472-35-0, NaH2PO4·2H2O
Supplier: Spectrum Chemical Mfg Cor S19301KG
Description
Spectrum™ Chemical Monobasic Sodium Phosphate, Dihydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 13472-35-0 | |
| 4.1 to 4.5 | |
| H6NaO6P | |
| O.O.[Na+].OP(O)([O-])=O | |
| 156.01 | |
| USP |
| 1 | |
| 1 kg | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| sodium dihydrate dihydrogen phosphate | |
| 98 to 103% | |
| Poly Bottle |
Chemical Identifiers
| 13472-35-0 | |
| 156.01 | |
| sodium dihydrate dihydrogen phosphate |
| H6NaO6P | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| O.O.[Na+].OP(O)([O-])=O |