missing translation for 'onlineSavingsMsg'
Learn More

Monobasic Sodium Phosphate, Dihydrate, USP, 98-103%, Spectrum™ Chemical

Catalog No. p-7157876
Change view
Click to view available options
Quantity:
500 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604662 500 g Poly Bottle
18604663 1 kg Poly Bottle
18604664 12 kg Poly Pail
18604665 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604662 Supplier Spectrum Chemical Mfg Cor Supplier No. S1930500GM
Only null left

S1930, 13472-35-0, NaH2PO4·2H2O

Spectrum™ Chemical Monobasic Sodium Phosphate, Dihydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.01
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
IUPAC Name sodium dihydrate dihydrogen phosphate
SMILES O.O.[Na+].OP(O)([O-])=O

Specifications

pH 4.1 to 4.5
CAS Max % 1
Quantity 500 g
Percent Purity 98 to 103%
Grade USP
Packaging Poly Bottle