missing translation for 'onlineSavingsMsg'
Learn More

Monobasic Sodium Phosphate, Anhydrous, USP, 98-103%, Spectrum™ Chemical

Catalog No. 18604661
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
25 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604661 25 kg Poly Bottle
18604658 2.5 kg Poly Bottle
18604659 12 kg Poly Bottle
18604660 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604661 Supplier Spectrum Chemical Mfg Cor Supplier No. SO18725KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO187, 7558-80-7, NaH2PO4

Spectrum™ Chemical Monobasic Sodium Phosphate, Anhydrous, USP is used as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
IUPAC Name sodium dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

pH 4.1 to 4.5
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 25 kg
Percent Purity 98 to 103%
Grade USP
Packaging Poly Bottle
Chemical Name or Material Monobasic Sodium Phosphate, Anhydrous