missing translation for 'onlineSavingsMsg'
Learn More

Monobasic Potassium Phosphate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015518
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015518 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015518 Supplier Merck Emd Millipore Supplier No. PHR13305G

Monobasic Potassium Phosphate is a water-soluble inorganic salt. It is used as a starting material in the synthesis of potassium metaphosphate.

Specifications

CAS 7778-77-0
Density 2.338 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula KH2 PO4
MDL Number MFCD00011401
Melting Point 252.6°C
Synonym Potassium phosphate monobasic; Monopotassium phosphate; Potassium dihydrogen phosphate; prim.-Potassium phosphate
Type Certified Reference Material
Quantity 5 g
Formula Weight 136.09
Grade Certified Reference Material
Show More Show Less