missing translation for 'onlineSavingsMsg'
Learn More

Monobasic Potassium Phosphate, Crystal, NF, 98-100.5%, Spectrum™ Chemical

Catalog No. 18604651
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604651 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604651 Supplier Spectrum Chemical Mfg Cor Supplier No. PO200500GM
Only null left

PO200, 7778-77-0, KH2PO4

Spectrum™ Chemical Monobasic Potassium Phosphate, Crystal, NF is used as a buffering agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 98 to 100.5%
Grade NF
Loss on Drying 0.01
Packaging Poly Bottle