missing translation for 'onlineSavingsMsg'
Learn More

mono-Methyl succinate, 95%

CAS: 3878-55-5 | C5H8O4 | 132.11 g/mol

Chemical Identifiers

CAS 3878-55-5
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.11
MDL Number MFCD00002788
InChI Key JDRMYOQETPMYQX-UHFFFAOYSA-N
Synonym monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
PubChem CID 77487
ChEBI CHEBI:75146
IUPAC Name 4-methoxy-4-oxobutanoic acid
SMILES COC(=O)CCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC127730100
View Documents Promotion Details
N/A 10 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3878-55-5
132.11
JDRMYOQETPMYQX-UHFFFAOYSA-N
77487
4-methoxy-4-oxobutanoic acid
C5H8O4
MFCD00002788
monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
CHEBI:75146
COC(=O)CCC(=O)O

Specifications

3878-55-5
100.0
White
Authentic
C5H8O4
MFCD00002788
monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
COC(=O)CCC(=O)O
132.11
CHEBI:75146
95%
mono-Methyl succinate, 98%
97.5
55°C to 59°C
151°C (20.0 mmHg)
94% min. (GC)
HO2CCH2CH2CO2CH3
02, 608
JDRMYOQETPMYQX-UHFFFAOYSA-N
4-methoxy-4-oxobutanoic acid
77487
132.11
Crystalline Chunks or Crystals
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-408-2

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only