missing translation for 'onlineSavingsMsg'
Learn More

Mono-6-O-(2-naphthyl)-per-O-methyl-alpha-cyclodextrin, TCI America™

Catalog No. M187610MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M187610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M187610MG Supplier TCI America Supplier No. M187610MG
Only null left

Chemical Identifiers

CAS 1019999-18-8
Molecular Formula C63H100O30
Molecular Weight (g/mol) 1337.463
InChI Key XRGIJNKZLAFOQY-ZOMSWCOISA-N
PubChem CID 121233863
SMILES COCC1C2C(C(C(O1)OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(O2)C(C7OC)OC)COC)COC)COC8=CC9=CC=CC=C9C=C8)COC)COC)OC)OC

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 1337.46
Physical Form Crystalline Powder
Chemical Name or Material Mono-6-O-(2-naphthyl)-per-O-methyl-alpha-cyclodextrin
TSCA No
Recommended Storage Refrigerator