accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1019999-18-8

CH3OOOOOCH3OCH3OOOCH3OCH3OOOCH3OH3COOOCH3OCH3OOOOH3COCH3OH3COCH3OOH3COCH3OH3COCH3

CAS RN 1019999-18-8

IUPAC Name: 31,32,33,34,35,36,37,38,39,40,41,42-dodecamethoxy-5,10,15,20,25-pentakis(methoxymethyl)-30-[(naphthalen-2-yloxy)methyl]-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane
Synonyms: 31,32,33,34,35,36,37,38,39,40,41,42-dodecamethoxy-5,10,15,20,25-pentakis(methoxymethyl)-30-[(naphthalen-2-yloxy)methyl]-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane
Molecular Weight (g/mol): 1337.46
Molecular Formula: C63H100O30
InChi Key: XRGIJNKZLAFOQY-UHFFFAOYNA-N
SMILES: COCC1OC2OC3C(COC)OC(OC4C(COC)OC(OC5C(COC6=CC=C7C=CC=CC7=C6)OC(OC6C(COC)OC(OC7C(COC)OC(OC1C(OC)C2OC)C(OC)C7OC)C(OC)C6OC)C(OC)C5OC)C(OC)C4OC)C(OC)C3OC
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.