missing translation for 'onlineSavingsMsg'
Learn More

Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate 95.0+%, TCI America™
GREENER_CHOICE

Catalog No. M28715ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28715ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M28715ML Supplier TCI America Supplier No. M28715ML

Chemical Identifiers

CAS 14802-03-0
Molecular Formula C16H35O3P
Molecular Weight (g/mol) 306.427
MDL Number MFCD00467188
InChI Key ZDFBXXSHBTVQMB-UHFFFAOYSA-N
Synonym (2-Ethylhexyl)phosphonic Acid Mono-2-ethylhexyl Ester, 2-Ethylhexyl Hydrogen (2-Ethylhexyl)phosphonate, HEHEHP, PC 88A
PubChem CID 85771
IUPAC Name 2-ethylhexoxy(2-ethylhexyl)phosphinic acid
SMILES CCCCC(CC)COP(=O)(CC(CC)CCCC)O

Specifications

Color Yellow
Quantity 5 mL
UN Number 3265
Formula Weight 306.43
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Mono-2-ethylhexyl (2-Ethylhexyl)phosphonate
EINECSNumber (2)-1961&(2)-2012
TSCA Yes