accessibility menu, dialog, popup

UserName

H3CCH3OPHOOCH3CH3

CAS RN 14802-03-0

IUPAC Name: (2-ethylhexyl)[(2-ethylhexyl)oxy]phosphinic acid
Synonyms: 2-ethylhexyl((2-ethylhexyl)oxy)phosphinic acid
Molecular Weight (g/mol): 306.43
Molecular Formula: C16H35O3P
InChi Key: ZDFBXXSHBTVQMB-UHFFFAOYNA-N
SMILES: CCCCC(CC)COP(O)(=O)CC(CC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results