missing translation for 'onlineSavingsMsg'
Learn More

mono-(2-Ethyl-5-hydroxyhexyl) phthalate, Mixture of diastereomers, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014150
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014150 Supplier Merck Emd Millipore Supplier No. 0917150MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 40321-99-1
Linear Formula C16H22O5
MDL Number MFCD01704544
Synonym 1,2-Benzenedicarboxylic acid 1-(2-ethyl-5-hydroxyhexyl) ester; Phthalic acid mono-(2-ethyl-5-hydroxyhexyl) ester
Type Analytical Standard
Quantity 50 mg
Formula Weight 294.34
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard