missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey Decorin ELISA Kit
ELISA
Supplier: Invitrogen™ EP22RB
Description
Monkey Decorin quantitates monkey Decorin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey Decorin.
Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.Chemical Identifiers
| bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; small leucine-rich protein 1 B; unnamed protein product | |
| 5 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Decorin | |
| 4 hrs 45 mins |
| 5 to 1,200 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; small leucine-rich protein 1 B; unnamed protein product | |
| 5 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 5 to 1,200 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Decorin | |
| 4 hrs 45 mins |