missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey Cathepsin S ELISA Kit
ELISA
$918.00 - $4130.00
Specifications
| Accession Number | Cat S; cathepsin S; Cats; CTSS |
|---|---|
| Assay Range | 28 to 7,000 pg/mL |
| Assay Sensitivity | 28 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Description
Monkey Cathepsin S quantitates monkey Cathepsin S in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey Cathepsin S.
The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.Specifications
| Cat S; cathepsin S; Cats; CTSS | |
| 28 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Cathepsin S | |
| 4 hrs 45 mins |
| 28 to 7,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |