missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey Calcitonin ELISA Kit
ELISA
Supplier: Invitrogen™ EEL214
Description
Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.Specifications
| 15.63 to 1000 pg/mL | |
| HRP | |
| ELISA Microplate (Dismountable) | |
| Plasma, Serum | |
| Colorimetric Microplate Reader | |
| <10 | |
| Biotin | |
| RUO | |
| Refer to product documentation for component specific storage temperature. | |
| 1 hr. 20 min. |
| 9.38 pg/mL | |
| Colorimetric | |
| ELISA | |
| Monkey | |
| <10 | |
| Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis | |
| 96 Tests | |
| Plasma: 100 μL, Serum: 100 μL | |
| Calcitonin | |
| 3 hr. 30 min. |