Learn More
Labconco Monitor Arm
Catalog No. 17200218
Change view
Click to view available options
For Use With (Equipment):
Enclosures
Catalog No. For Use With (Equipment)
17200218 Enclosures
1 options
Catalog No. 17200218 Supplier Labconco Supplier No. 3997300
Desk Monitor Arm allows independent placement a computer monitor or tablet for ease of use while working in a enclosure.
  • Desk Monitor Arm for use with Labconco PC Mount kit and Labconco base stands only
  • Polished Aluminum
  • Allows independent placement of a monitor or tablet (tablet case not included)
  • Capacity 7 to 25 lbs. (3.2 to 11.3 kg)
  • PC Mount kit required and sold separately
  • Installation required
Specifications
For Use With (Equipment) Enclosures
Product Type Monitor Arm