missing translation for 'onlineSavingsMsg'
Learn More

Monensin sodium salt, 90%, Thermo Scientific Chemicals

Catalog No. AC461450010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461450010 1 g
AC461450050 5 g
AC461450250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC461450010 Supplier Thermo Scientific Chemicals Supplier No. 461450010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22373-78-0
Molecular Formula C36H61NaO11
Molecular Weight (g/mol) 692.86
InChI Key XOIQMTLWECTKJL-FBZUZRIGSA-M
IUPAC Name sodium (2S,3R,4S)-4-[(2S,5R,7S,8R,9S)-2-[(2S,2'R,3'S,5R,5'R)-2-ethyl-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3'-methyl-[2,2'-bioxolan]-5-yl]-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl]-3-methoxy-2-methylpentanoate
SMILES [Na+].CC[C@]1(CC[C@@H](O1)[C@]1(C)CC[C@]2(C[C@H](O)[C@@H](C)[C@H](O2)[C@@H](C)[C@@H](OC)[C@H](C)C([O-])=O)O1)[C@@H]1O[C@H](C[C@@H]1C)[C@H]1O[C@@](O)(CO)[C@H](C)C[C@@H]1C

Specifications

Melting Point 267.0°C to 269.0°C
Color Clear Colorless to Light Yellow
Assay Percent Range 88 % min. (HPLC)
Quantity 1 g
Formula Weight 692.85
Chemical Name or Material Monensin sodium salt
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Fatal if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Rinse mouth.

EINECSNumber 244-941-7

RUO – Research Use Only