accessibility menu, dialog, popup

UserName

NaCH3(S)(R)O(S)H3C(R)(S)OH(R)CH3(S)O(S)H3C(R)OCH3(S)H3COOO(R)O(R)(S)CH3(S)O(R)OHOH(R)H3C(S)CH3

CAS RN 22373-78-0

IUPAC Name: sodium (2S,3R,4S)-4-[(2S,5R,7S,8R,9S)-2-[(2S,2'R,3'S,5R,5'R)-2-ethyl-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3'-methyl-[2,2'-bioxolan]-5-yl]-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl]-3-methoxy-2-methylpentanoate
Synonyms: sodium (2S,3R,4S)-4-[(2S,5R,7S,8R,9S)-2-[(2S,2'R,3'S,5R,5'R)-2-ethyl-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3'-methyl-[2,2'-bioxolan]-5-yl]-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl]-3-methoxy-2-methylpentanoate
Molecular Weight (g/mol): 692.86
Molecular Formula: C36H61NaO11
InChi Key: XOIQMTLWECTKJL-FBZUZRIGSA-M
SMILES: [Na+].CC[C@]1(CC[C@@H](O1)[C@]1(C)CC[C@]2(C[C@H](O)[C@@H](C)[C@H](O2)[C@@H](C)[C@@H](OC)[C@H](C)C([O-])=O)O1)[C@@H]1O[C@H](C[C@@H]1C)[C@H]1O[C@@](O)(CO)[C@H](C)C[C@@H]1C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)