missing translation for 'onlineSavingsMsg'
Learn More

Monensin sodium salt, 90% min., Thermo Scientific Chemicals

Catalog No. AAJ6166903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6166903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6166903 Supplier Thermo Scientific Chemicals Supplier No. J6166903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Monensin sodium salt is used in potentiometric and spectroscopic studies of alkali metal ion complexes. Monensin (Na) is a polyether small molecule ionophore, capable of forming stable complexes to monovalent cations with specific affinity for Na+. Monensin presents a highly lipophilic scaffold around the bound ion, allowing movement of the complex through the lipid bilayer and consequent transport of the ion out of the cell. Excessive flux of ions out of the cell produces a cytotoxic effect and generates the antibiotic properties of Monensin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Monensin sodium salt is used in potentiometric and spectroscopic studies of alkali metal ion complexes. Monensin (Na) is a polyether small molecule ionophore, capable of forming stable complexes to monovalent cations with specific affinity for Na+. Monensin presents a highly lipophilic scaffold around the bound ion, allowing movement of the complex through the lipid bilayer and consequent transport of the ion out of the cell. Excessive flux of ions out of the cell produces a cytotoxic effect and generates the antibiotic properties of Monensin.

Solubility
Soluble in methanol at 50mg/ml

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 22373-78-0
Molecular Formula C36H61NaO11
Molecular Weight (g/mol) 692.863
MDL Number MFCD00077826
InChI Key XOIQMTLWECTKJL-JXDONITHSA-M
Synonym monensin, sodium salt, high purity, monensin sodium salt, monensin, monosodium salt, 4.5 dec-7-yl 3s,4r-3-methoxy-2-me thylpentanoic acid, sodium salt, 4-2-5-3r-5-3s,5r,6r-6-hydroxy-6-hydroxymethyl-3,5-dimethyl 2h-3,4,5, 6-tetrahydropyran-2-yl-3-methyloxolan-2-yl 5s-5-ethyloxolan-2-yl 5s,8s,2r ,9r-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro, 4-2-2-ethyl-5'-6-hydroxy-6-hydroxymethyl-3,5-dimethyltetrahydro-2h-pyran-2-yl-3'-methyloctahydro-2,2'-bifuran-5-yl-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro 4.5 decane-7-yl-3-methoxy-2-methylpentanoic acid sodium salt, potassium 4-2-5-ethyl-5-5-6-hydroxy-6-hydroxymethyl-3,5-dimethyloxan-2-yl-3-methyloxolan-2-yl oxolan-2-yl-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro 4.5 decan-7-yl-3-methoxy-2-methylpentanoate
PubChem CID 133612076
IUPAC Name sodium;(3S,4R)-4-[(2R,5S,7R,8S,9R)-2-[(2S,5R)-5-ethyl-5-[(2S,3R,5S)-5-[(2R,3R,5S,6S)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]oxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]-3-methoxy-2-methylpentanoate
SMILES CCC1(CCC(O1)C2(CCC3(O2)CC(C(C(O3)C(C)C(C(C)C(=O)[O-])OC)C)O)C)C4C(CC(O4)C5C(CC(C(O5)(CO)O)C)C)C.[Na+]

Specifications

Melting Point 267°C to 269°C
Color White
Quantity 1 g
UN Number UN3462
Beilstein 4122200
Merck Index 14,6246
Solubility Information Soluble in methanol at 50mg/ml
Formula Weight 692.86
Percent Purity 90 to 95.5%
Physical Form Powder
Chemical Name or Material Monensin sodium salt
Show More Show Less
Hazard Category H300
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
EINECSNumber 244-941-7
RTECSNumber JH2830000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only