missing translation for 'onlineSavingsMsg'
Learn More

Monastrol, 98%, Thermo Scientific Chemicals

Catalog No. AAJ67096LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67096LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67096LB0 Supplier Thermo Scientific Chemicals Supplier No. J67096LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 329689-23-8
Molecular Formula C14H16N2O3S
Molecular Weight (g/mol) 292.353
MDL Number MFCD00813077
InChI Key LOBCDGHHHHGHFA-UHFFFAOYSA-N
Synonym monastrol, ethyl 4-3-hydroxyphenyl-6-methyl-2-thioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, 4-3-hydroxyphenyl-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4h-pyrimidin-5-carboxylic acid ethyl ester, d0zk2g, monastrol hplc , solid, inverted exclamation marka-monastrol, 6-methyl-4-3-hydroxyphenyl-2-thioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, ethyl 6-3-hydroxyphenyl-2-mercapto-4-methyl-1,6-dihydropyrimidine-5-carboxylate, ethyl 6-3-hydroxyphenyl-4-methyl-2-sulfanyl-1,6-dihydropyrimidine-5-carboxylate, ethyl 6-3-hydroxyphenyl-4-methyl-2-thioxo-1,3,6-trihydropyrimidine-5-carboxy late
PubChem CID 2987927
ChEBI CHEBI:75382
IUPAC Name ethyl 4-(3-hydroxyphenyl)-6-methyl-2-sulfanylidene-3,4-dihydro-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=C(NC(=S)NC1C2=CC(=CC=C2)O)C

Specifications

Melting Point 179°C to 180°C
Color White to Yellow
Quantity 5 mg
Sensitivity Light sensitive
Merck Index 14,6244
Formula Weight 292.36
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Monastrol
RTECSNumber UV6262400
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only