missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdic Acid 85%, ACS Reagent, Reagents
Molybdic Acid 85%, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1109000500C2
Specifications
| Molybdic Acid 85%, ACS Reagent | |
| 7782-91-4 | |
| 500 g | |
| Ammonium Molybdate | |
| O[Mo](O)(=O)=O | |
| 161.96 | |
| Glass Bottle |
| White or Slightly Yellow | |
| Solid | |
| H2MoO4 | |
| VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
| dioxomolybdenumdiol | |
| ACS | |
| 3.1 |
Chemical Identifiers
| 7782-91-4 | |
| 161.96 | |
| Ammonium Molybdate | |
| O[Mo](O)(=O)=O |
| H2MoO4 | |
| VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
| dioxomolybdenumdiol |