missing translation for 'onlineSavingsMsg'
Learn More

Molybdic Acid 85%, ACS Reagent, Reagents

Catalog No. p-200021173
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338172 100 g
01338171 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338172 Supplier Labchem Inc Supplier No. C1109000100C2
Only null left

Molybdic Acid 85%, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-91-4
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym Ammonium Molybdate
IUPAC Name dioxomolybdenumdiol
SMILES O[Mo](O)(=O)=O

Specifications

Color White or Slightly Yellow
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.1
Chemical Name or Material Molybdic Acid 85%, ACS Reagent