missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) Oxide, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1004030100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1004030100 100 g Plastic bottle
M1004030500 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1004030100 Supplier Technidata Supplier No. 1.00403.0100
For analysis

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Physical Form Solid
Quantity 100 g
Formula Weight 143.94
Odor Odorless
Packaging Plastic bottle
Appearance Yellow solid
Chemical Name or Material Molybdenum(VI) oxide
Hazard Category H351: Suspected of causing cancer.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
EINECSNumber 215-204-7