Learn More
Molybdenum(VI) oxide, ACS, 99.5% min
Description
Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in oil industr. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Source of molybdenum compounds, in agriculture, in analytical chemistry, as a corrosion inhibitor, in ceramic glazes, enamels and pigments, as a catalyst
Specifications
Specifications
| Boiling Point | 1155°C (sublimation) |
| Melting Point | 795°C |
| Color | White to Gray |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99.5% |
| UN Number | UN3288 |
| Merck Index | 14,6239 |
| Solubility Information | Slightly soluble in water; Soluble in concentrated mineral acids,alkali hydroxide solutions. |
| Formula Weight | 143.94 |
| Show More |
Safety and Handling
| Hazard Category | H319-H335-H351 |
| Hazard Statement | GHS H Statement H300-H351-H319-H335 Fatal if swallowed. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 215-204-7 |
| RTECSNumber | QA4725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only