missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, ACS, 99.5% min

Catalog No. AA3668722
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3668722 100 g
AA3668736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3668722 Supplier Thermo Scientific Chemicals Supplier No. 03668722

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in oil industr. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Source of molybdenum compounds, in agriculture, in analytical chemistry, as a corrosion inhibitor, in ceramic glazes, enamels and pigments, as a catalyst

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155°C (sublimation)
Melting Point 795°C
Color White to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5%
UN Number UN3288
Merck Index 14,6239
Solubility Information Slightly soluble in water; Soluble in concentrated mineral acids,alkali hydroxide solutions.
Formula Weight 143.94
Percent Purity ≤99.5%
Grade ACS
Odor Odorless
Density 4.692 g/mL
Chemical Name or Material Molybdenum(VI) oxide, For analysis ACS
Show More Show Less
Hazard Category H319-H335-H351
Hazard Statement GHS H Statement
H300-H351-H319-H335
Fatal if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 215-204-7
RTECSNumber QA4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only