Learn More
Molybdenum(VI) oxide, 99.95% (metals basis)
Description
Molybdenum(VI) oxide is a very promising and stable molybdenum oxide for the electrochromic applications and used as a catalyst in oil industries. It is also used to make metal molybdenum. It has a wide band gap n-type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties. It has been widely investigated as a smart material for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials. Molybdenum(VI) oxide (MoO3) is employed as an adhesive between enamels and metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1155°C (sublimation) |
| Melting Point | 795°C |
| Color | White |
| Physical Form | Sublimed Powder |
| Quantity | 100 g |
| Assay Percent Range | 99.95% (metals basis) |
| Solubility Information | Barely soluble in water; soluble in base. |
| Formula Weight | 143.94 |
| Odor | Odorless |
| Appearance | White to gray powder |
| Show More |
Safety and Handling
| Hazard Category | H319-H335-H351 |
| Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 215-204-7 |
| RTECSNumber | QA4725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only