missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, 99.95% (metals basis)

Catalog No. AA1183722
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1183722 100 g
AA1183736 500 g
AA11837A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1183722 Supplier Thermo Scientific Chemicals Supplier No. 01183722

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

Molybdenum(VI) oxide is a very promising and stable molybdenum oxide for the electrochromic applications and used as a catalyst in oil industries. It is also used to make metal molybdenum. It has a wide band gap n-type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties. It has been widely investigated as a smart material for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials. Molybdenum(VI) oxide (MoO3) is employed as an adhesive between enamels and metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155°C (sublimation)
Melting Point 795°C
Color White
Physical Form Sublimed Powder
Quantity 100 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Barely soluble in water; soluble in base.
Formula Weight 143.94
Odor Odorless
Appearance White to gray powder
Density 4.692 g/cm3 at 20°C
Chemical Name or Material Molybdenum(VI) oxide
Show More Show Less
Hazard Category H319-H335-H351
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 215-204-7
RTECSNumber QA4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only