missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, 99.5+%, ACS reagent

Catalog No. AC447401000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447401000 100 g Plastic bottle
AC447405000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447401000 Supplier Thermo Scientific Chemicals Supplier No. 447401000

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155.0°C
Melting Point 795.0°C
Quantity 100 g
Linear Formula MoO3
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in conc. mineral acids, sol. of alkali, hydroxides and ammonia, soluble in potassium bitartrate
Formula Weight 143.94
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Molybdenum(VI) oxide

RUO – Research Use Only