missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

Catalog No. AA40866KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40866KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40866KS Supplier Thermo Scientific Chemicals Supplier No. 040866KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7439-98-7 | Mo | 95.95 g/mol

Molybdenum sputtering target is used as resistance heating elements in furnaces, as filament supports in electric lamps, and as electrodes for mercury vapor lamps. It is also used as an alloying agent in certain grades of steel, Permalloys and Stellites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 1 Ea.
Solubility Information Insoluble in water. Soluble in nitric acid and sulfuric acid. Slightly soluble in hydrochloric acid.
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Molybdenum sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only