missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum, plasma standard solution, Specpure™ Mo 1000μg/mL

CAS: 7439-98-7 | Mo | 95.95 g/mol

Supplier:  Thermo Scientific Chemicals 035758AE

 View more versions of this product

Catalog No. AA35758AE


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Molybdenum
7439-98-7
Liquid
MFCD00003465
[Mo]
95.95
23932
Mo 1000μg/mL
23 hPa (17mm Hg) at 20°C
Mo
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
100 mL
CHEBI:28685

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
CHEBI:28685
[Mo]
Mo
MFCD00003465
23932
molybdenum
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8; Packing Group: III

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only