missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum ICP-MS Standard, 1000 ppm Mo in 3% NH4OH, Ricca Chemical
Supplier: Ricca Chemical Company MSMO1KW100
Specifications
| Molybdenum ICP-MS Standard | |
| 13106-76-8,1336-21-6,7732-18-5 | |
| 1,000ppm Mo | |
| 1 mL = 1 mg Mo | |
| Colorless to Green/Yellow | |
| 100°C | |
| Miscible with water | |
| ICP Single-element Standard Solution | |
| Laboratory |
| Natural LDPE Bottle | |
| 95.0,2.85,0.19 | |
| 99.0,2.96,0.2 | |
| Liquid | |
| 0.0°C | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| 100 mL |
Chemical Identifiers
| 13106-76-8 |