missing translation for 'onlineSavingsMsg'
Learn More

Molybdate Reagent, Taylor Type, 1.2% (w/v) Sodium Molybdate in 4.9% (v/v) Sulfuric Acid, Ricca Chemical

Supplier:  Ricca Chemical Company RI51905

Catalog No. 51905

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Molybdate Reagent
10102-40-6 , 7732-18-5 , 7664-93-9
1.11, 93.0, 8.34
MFCD00149170
TVXXNOYZHKPKGW-UHFFFAOYSA-N
disodium dioxomolybdenumbis(olate)
16211258  
Cubitainer
20 L
Taylor Type
1.07, 89.0, 8.02
MoNa2O4
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
[Na+].[Na+].[O-][Mo]([O-])(=O)=O
205.93
CHEBI:75213
4.9% (v/v) Sulfuric Acid
1.2% (w/v) Sodium Molybdate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions