missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. pellets

Catalog No. AA33551A7
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33551A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA33551A7 Supplier Thermo Scientific Chemicals Supplier No. 033551A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol

Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. Pellets is used as an sorbent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Color Brown
Quantity 5 kg
Sensitivity Hygroscopic
Physical Form Pellet
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves 5A, 3 to 4mm (0.12 to 0.16 in.) dia. pellets
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only