missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 5Å, Solstice

Catalog No. 60045155
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with Inliner
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045155 5 kg Drum with Inliner
60045154 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045155 Supplier Solstice Supplier No. 2086205KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Beads, 8-12 mesh

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Pore Size 5 Å
Quantity 5 kg
Mesh Size 8 to 12mesh
Packaging Drum with Inliner
Physical Form Beads
Linear Formula Ca/nNa12-2n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name aluminum;sodium;dioxido(oxo)silane
Chemical Name or Material Molecular Sieves, 5Å