missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 5Å, Solstice

Catalog No. 60045154
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
5 kg
Packaging:
Drum with Inliner
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045154 1 kg Poly Bottle
60045155 5 kg Drum with Inliner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 60045154 missing translation for 'mfr' Solstice missing translation for 'supplierNo' 2086201KG

Beads, 8-12 mesh

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Pore Size 5 Å
Quantity 1 kg
Packaging Poly Bottle
Mesh Size 8 to 12mesh
Physical Form Beads
Linear Formula Ca/nNa12-2n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name Molecular sieves 5A
Chemical Name or Material Molecular Sieves, 5Å