missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 3A, Pellets, Dia.: 1.6mm, Honeywell Fluka™
Pellets, 1.6 mm
$3,078.29
Chemical Identifiers
| CAS | 308080-99-1 |
|---|---|
| Molecular Formula | AlNaO6Si2 |
| Molecular Weight (g/mol) | 202.135 |
| MDL Number | MFCD00131613 |
| InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
| PubChem CID | 19758701 |
| SMILES | [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Price | Quantity & Availability | |||||
|
60046245
|
Honeywell-Fluka
3342865KG |
5 kg |
Each for $3,078.29
|
|
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 308080-99-1 | |
| 202.135 | |
| URGAHOPLAPQHLN-UHFFFAOYSA-N | |
| [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
| AlNaO6Si2 | |
| MFCD00131613 | |
| 19758701 |
Specifications
| Dia.: 1.6mm | |
| Poly Bottle | |
| 308080-99-1 | |
| AlNaO6Si2 | |
| MFCD00131613 | |
| URGAHOPLAPQHLN-UHFFFAOYSA-N | |
| Molecular sieves 3A | |
| 19758701 |
| 2g/cm3 | |
| Pellets | |
| 400°C | |
| KnNa12-n[(AlO2)12(SiO2)12] · xH2O | |
| NONH for all modes of transport | |
| [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] | |
| 202.135 | |
| Molecular sieves, UOP Type 3Å |