missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol

Supplier:  Thermo Scientific Chemicals L053350I

Quantity 5000 g

 View more versions of this product

Catalog No. AAL053350I


Description

Description

Molecular sieves, 3A is used as a desiccant in petroleum and chemical industries. It is useful for the drying of unsaturated hydrocarbons such as ethylene, propylene and butadiene. It is also used in the removal of water from ethanol. It finds application in the drying of certain chemicals namely ethanol, refrigerants and natural gas. It plays an important role in selective adsorption of water in insulated glass (IG) and polyurethane. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

308080-99-1
202.135
URGAHOPLAPQHLN-UHFFFAOYSA-N
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
AlNaO6Si2
MFCD00131613
19758701

Specifications

Molecular sieves, 3A
Hygroscopic
308080-99-1
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
19758701
5000 g
1 to 2mm (0.04 to 0.08 in) beads
AlNaO6Si2
Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone.
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
202.135
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions